KIMBLE® 105° Bend Vacuum Take-Off Adapters, Outer Hose Connection, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
205600-1420 205600-1922
KT205600-1420EA 226.14 CAD
KT205600-1420 KT205600-1922 KT205600-2440
KIMBLE® 105° Bend Vacuum Take-Off Adapters, Outer Hose Connection, DWK Life Sciences
Adapters Adapters, Joints with Tube Connectors
Adapters are bent at a 105° angle for use in vacuum distillation.
  • Top [ST] outer joint
  • Bottom [ST] inner joint
  • Both joints are the same size
  • With hose connection and drip tip
Manufactured from 33 expansion, low extractable borosilicate glass.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
SPECIFICATIONS
Environmentally Preferable