Cubis® II Essential Premium Micro Balances, MCE Series, Standard Versions, Sartorius
Supplier: Sartorius
MCE6.6S2S00F MCE3.6P2S00F MCE10.6S2S00M MCE6.6S2S00M CA76334-114 MCE3.6P-M-MTS MCE10.6S2S00F
CA76334-104EA 51970.55 CAD
CA76334-104 CA76334-106 CA76334-110 CA76334-112 CA76334-114 CA76334-154 CA76334-274
Cubis® II Essential Premium Micro Balances, MCE Series, Standard Versions, Sartorius
Balances and Scales
These Cubis® II premium MCE series - essential user interface balances are ideal for high-performance portfolio of both labs weighing hardware and software.
  • Individually configured from different components
  • Fully customizable hardware, software, and connectivity
  • Customizable modularity, intelligence by design, leading Performance, high speed accuracy, error-free operations, full end to end data integrity, reliable support
  • Easy to use manual glass draft shield, safety data transfer
  • Removable flat stainless-steel draft shield frame
  • Large 7" color touch display including touch, scroll, swipe functionality
  • Diverse QApp packages for optional software extension
  • Automatic, motorized leveling, gesture control, climate monitoring, built-in ionizer for electrostatic elimination
These balances offer modern user interfaces, pharmaceutical and GxP compliance, including data handling, data integrity and connectivity, ergonomic sample handling, easy process integration, and unlimited communication at the highest level of accuracy and precision.
The status center shows all information about your balance and environmental conditions. Automatically execute time-controlled actions for backup.
The Cubis® II balance contains an 'Alibi Memory'. The system automatically stores weighing data in a ring buffer. An electronic signature can be used to sign the final report for weighing processes. Based on a secure password, the combination of username and password is fully compliant with 21CFR.
Different draft shields and sereis are:
F - Filter draft shield micro balances
M - Motorized draft shield micro balances.
Certifications: All requirements for FDA directive 21 CFR part 11 and EU Annex 11 compliance.
Delivery information: There are different optional software packages, each contains applications or functions to enhance and customize the balance for the user's workflows. choice of QApp package: pharma; advanced applications: utilities; connectivity or the all-inclusive QApps package includes all of these four packages. (denoted by QP99).