Cubis® II Advanced Premium Micro Balances, MCA Series, Standard Versions, Sartorius
Supplier: Sartorius
Cubis® II
MCA3.6PM-S00 MCA10.6S-2S00M MCA6.6S-2S00-M MCA6.6S-2S00-F MCA6.6S-2S00-F MCA3.6P-2S00-M MCA10.6S-M-MTS MCA6.6SF-S00 MCA3.6P-2S00-F MCA6.6S-2S00-M MCA3.6P-2S00-M MCA3.6PM-S00 MCA6.6SM-S00 MCA3.6P-2S00-F MCA10.6S-2S00F MCA6.6SM-S00 MCA10.6S-2S00M MCA10.6SM-S00 MCA6.6SF-S00 MCA10.6S-2S00F
CA76333-952EA 68189.75 CAD
CA76333-952 76377-374 CA76334-220 76377-362 CA76377-374 76377-372 76377-394 CA76377-364 76377-382 CA76377-376 CA76334-224 CA76334-222 76377-390 76377-358 76377-368 CA76333-954 76377-376 76377-364 CA76333-956 CA76377-394
Cubis® II Advanced Premium Micro Balances, MCA Series, Standard Versions, Sartorius
Balances and Scales
These Cubis® II premium MCA series - advanced user interface balances are ideal for high-performance portfolio of both labs weighing hardware and software.
  • Individually configured from different components
  • Fully customizable hardware, software, and connectivity
  • Customizable modularity, intelligence by design, leading Performance, high speed accuracy, error-free operations, full end to end data integrity, reliable support
  • Easy to use manual glass draft shield, safety data transfer
  • Removable flat stainless-steel draft shield frame
  • Large 7" color touch display including touch, scroll, swipe functionality
  • Diverse QApp packages for optional software extension
  • Automatic, motorized leveling, gesture control, climate monitoring, built-in ionizer for electrostatic elimination
These balances offer modern user interfaces, pharmaceutical and GxP compliance, including data handling, data integrity and connectivity, ergonomic sample handling, easy process integration, and unlimited communication at the highest level of accuracy and precision.
The status center shows all information about your balance and environmental conditions. Automatically execute time-controlled actions for backup.
The Cubis® II balance contains an 'Alibi Memory'. The system automatically stores weighing data in a ring buffer. An electronic signature can be used to sign the final report for weighing processes. Based on a secure password, the combination of username and password is fully compliant with 21CFR.
Different draft shields and series are:
F - Filter draft shield micro balances
M - Motorized draft shield micro balances
Certifications: All requirements for FDA directive 21 CFR part 11 and EU Annex 11 compliance.
Delivery information: There are different optional software packages, each contains applications or functions to enhance and customize the balance for the user's workflows. choice of QApp package: pharma; advanced applications: utilities; connectivity or the all-inclusive QApps package includes all of these four packages. (denoted by QP99).