KIMBLE® KONTES® Funnel Condenser For Sublimation Assembly, DWK Lifesciences
Supplier: DWK Life Sciences
KIMBLE®
746125-0001
KT746125-0001EA 38.27 CAD
KT746125-0001
KIMBLE® KONTES® Funnel Condenser For Sublimation Assembly, DWK Lifesciences
Condensers
Sublimation assembly with vertically adjustable condenser tube for use with microscale vessels.
  • Condenser has vertically adjustable tube
  • Adapter has 5 to 8 mm ID range
  • Compression cap and FKM O-ring provide a vacuum-tight seal and allow for adjustment
Manufactured from 33 expansion, low extractable borosilicate glass.
Certifications: Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Delivery information: Consists of funnel condenser and inlet adapter with cap and O-ring.