KEM-KIT® Macroscale Standard Taper Kit, [ST] 19/22, DWK Life Scineces
Supplier: DWK Life Sciences
KIMBLE®
269800-0000
KT269800-0000EA 1904.4 CAD
KT269800-0000
KEM-KIT® Macroscale Standard Taper Kit, [ST] 19/22, DWK Life Scineces
Microchemistry Kits
KEM-KITS® contain a selection of basic small scale glassware for extraction, reflux, reflux with addition, fractionation, and distillation (steam, atmospheric and vacuum), as well as for other organic preparations.
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Delivery information: Supplied in a double-wall heavy-duty polyethylene storage case with a handle and hinged lid.