Drying Tube, Bent, DWK Life Sciences
Supplier: DWK Life Sciences
746095-0000
KT746095-0000CS 102.16 CAD
KT746095-0000
Drying Tube, Bent, DWK Life Sciences
Tubes Drying Tubes
This drying tube is used with an unsealed system to protect moisture-sensitive reactions from the atmosphere.
  • Modified drying tube with an expansion bulb for CaCl₂
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.