Blackout Paper Labels
Supplier: GA INTERNATIONAL INC
807656032054 807656032061 807656042503 807656032047 807656022307 807656104843 807656104836 807656022338 807656022345 807656022352 807656022369 807656022291
76210-158RL 351.82 CAD
76210-158 76210-288 76210-290 76210-292 76274-302 76274-304 76274-306 76274-308 76274-310 76274-312 76274-314 76274-756
Blackout Paper Labels
Labels
These permanent blackout labels will conceal any existing printed data, including text and barcodes. Ideally suited for general use applications in labs and offices. These cover-up labels are compatible with all standard thermal transfer barcode printers and can be printed with readable text and barcodes.
  • Temperature range: −54 to +93 °C (−65.2 to 199.4 °F)
  • Unique opaque design allows the labels to block-out all pre-existing information
  • For general office, laboratory, and warehouse applications
  • Compatible with all major thermal transfer printer brands