Cryogenic Labels for Direct Thermal Barcode Printers
Supplier: GA INTERNATIONAL INC
807656032528 807656032504 807656032481 807656032511 807656032498 807656022864 807656022871 807656005379 807656022888 807656006765 807656006604 807656022680 807656022697 807656022703 807656022710 807656022727 807656022734 807656022741 807656022758 807656022765 807656022772 807656022789 807656022802 807656022819 807656028712 807656028729 807656022826 807656022833 807656022840 807656022857 807656006109 807656022895 807656022901 807656022918 807656022925 807656022932 807656005973 807656005737 807656022949 807656022956 807656022963 807656022970 807656022987 807656022994 807656023007 807656023014 807656023021 807656023038 807656028736 807656028743 807656006017 807656023069 807656023076 807656023083 807656023090 807656023106 807656023113 807656023120 807656023137 807656023144 807656023151 807656023199 807656028552 807656028569 807656028576 807656028583 807656028590 807656028606 807656028620 807656028637 807656028644 807656023212 807656023229 807656109824 807656109831 807656111513 807656111711 807656060583 807656060606 807656060613 807656060620 807656060637 807656028613 807656032429 807656032436 807656101460 807656101804 807656102276 807656102283 807657612941
76210-164RL 381.84 CAD
76210-164 76210-166 76210-168 76210-170 76210-172 76210-174 76274-638 76274-640 76274-642 76274-644 76274-946 76274-948 76274-950 76274-952 76274-954 76274-956 76274-966 76274-968 76274-970 76274-972 76275-030 76275-032 76275-034 76275-038 76275-040 76275-042 76275-044 76275-046 76275-048 76275-050 76275-052 76275-054 76275-056 76275-058 76275-060 76275-062 76275-064 76275-066 76275-068 76275-070 76275-072 76275-074 76275-076 76275-078 76275-080 76275-082 76275-084 76275-086 76275-088 76275-090 76275-092 76275-094 76275-096 76275-098 76275-100 76275-104 76275-106 76275-108 76275-110 76275-112 76275-114 76275-116 76275-118 76275-120 76275-122 76275-124 76275-126 76275-128 76275-130 76275-132 76275-134 76275-136 76275-138 76275-140 76275-142 76275-144 76275-146 76275-148 76275-150 76275-152 77320-988 77320-990 77320-992 77320-994 77320-966 77320-968 77320-970 77320-972 77320-974 77320-908 77320-910 77320-912 77320-978 77320-980 77320-982 77320-984 77321-064
Cryogenic Labels for Direct Thermal Barcode Printers
Labels
Permanent direct thermal cryo labels for vials, microtubes, microplates, freezer boxes, and plastic bags. These barcode labels withstand long-term storage in cryogenic conditions.
  • Temperature range: –196 to 93 ℃ (–321 to 199 ℉)
  • Labels can withstand long-term storage in liquid Nitrogen (–196 ℃), laboratory freezers (–80 ℃, –20 ℃), and transportation in dry ice
  • Ideal for the identification of vials, microtubes, microplates, and freezer boxes
  • Printable with all major direct thermal printer brands
  • No need for extra lamination
These cryogenic barcode labels are specially designed for printing with direct thermal printers. The thermoplastic waterproof labels are specifically designed to permanently adhere to vials, microcentrifuge tubes, microtubes, microplates, glass tubes, freezer boxes, plastic bags, and other specimen containers that will undergo long-term cryogenic and deep-freeze storage.
These labels have a strong affinity for glass, polypropylene, polyethylene, and other plastic surfaces. The cryogenic labels will remain adhered to tubes through multiple freeze-thaw cycles without cracking or peeling off. These direct thermal cryogenic labels are compatible with most cryogenic storage systems without any hindrance or risk of being scraped off. These labels do not require a self-laminate to work.