KIMBLE® Barbed Hose Connectors, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
893500-0022 893750-0021 893750-0022
KT893500-0022EA 74.79 CAD
KT893500-0022 KT893750-0021 KT893750-0022
KIMBLE® Barbed Hose Connectors, DWK Life Sciences
Fittings and Connectors
These hose connectors are useful in constructing manifolds or connecting apparatus.
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
SPECIFICATIONS
Environmentally Preferable