Permanent Autoclave Resistant Thermal Transfer Labels
Supplier: GA INTERNATIONAL INC
807656031699 807656031705 807656031712 807656031682 807656020204 807656020211 807656020228 807656028040 807656019918 807656019925 807656019956 807656019963 807656019970 807656019987 807656019994 807656020006 807656020037 807656020105 807656020174 807656020181 807656028026 807656028033 807656020198 807656031859
76210-240RL 188.51 CAD
76210-240 76210-242 76210-244 76210-480 76273-518 76273-520 76273-522 76273-524 76274-560 76274-562 76274-568 76274-570 76274-572 76274-574 76274-576 76274-578 76274-584 76274-598 76274-612 76274-614 76274-616 76274-618 76274-620 76274-622
Permanent Autoclave Resistant Thermal Transfer Labels
Labels
Permanent autoclave-resistant labels withstand various sterilization procedures, including in steam autoclaves, dry heat ovens, gamma irradiators, and ethylene oxide gas sterilizers.
  • Temperature range: –60 to +150 °C (–76 to +302 °F)
  • Ideal for use in repeated steam autoclave sterilization cycles, gamma irradiation (up to 50 KGy), and ethylene oxide sterilization
  • Versatile labels adhere to a variety of surfaces including metal, plastic, and glass
  • Printable with all major thermal transfer printer brands
  • Waterproof and weatherproof
  • RR-class resin ribbon recommended
High temperature-resistant permanent labels for identifying various sterilization containers and surfaces that will undergo autoclaving and dry heat sterilization. Temperature ranges from –60 to +150 °C (–76 to +302 °F). The labels can be applied to metal, plastic, and glass surfaces, and will withstand up to 20 autoclave sterilization cycles at 121 °C (250 °F) and 16.5 psi pressure.
The labels and the printout can withstand high temperature exposure, as well as gamma irradiation (up to 50 KGy) and ethylene oxide sterilization. Ideal for identifying containers and surfaces that need to be sterilized inside autoclaves, dry heat sterilizing equipment, and with gamma irradiators. Waterproof, weatherproof, thermal transfer printable.