KIMBLE® Magnetic Stir Bars, Cross-shaped, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
791190-0100 791190-0114
10842-774CS 39.18 CAD
10842-774 10842-776
KIMBLE® Magnetic Stir Bars, Cross-shaped, DWK Life Sciences
Stirrers Magnetic Stirring Bars
This all-purpose stir bar permits stirring in closed containers without contamination.
  • PTFE-covered X-shaped magnetic stirring bar
  • More turbulence with less vibration than straight stir bars
  • Autoclavable
Ideal for use with conical bottom HPLC reservoirs.