KIMBLE® NMR Sample Tubes, 3 mm, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
897800-0000 897805-0000 897820-0008 897830-0000 897835-0000 897840-0000 897840-0008
KT897800-0000CS 226.88 CAD
KT897800-0000 KT897800-0008 KT897805-0000 KT897820-0008 KT897825-0000 KT897830-0000 KT897835-0000 KT897835-0008 KT897840-0000 KT897840-0008
KIMBLE® NMR Sample Tubes, 3 mm, DWK Life Sciences
Tubes NMR Tubes
Designed for high resolution work with NMR spectrometers.
  • High-quality tubes are ideal for sealing directly to vacuum manifolds, joints, or valves
  • Tubes feature a sandblasted marking spot
Manufactured from ASTM Type 1 borosilicate glass under rigid quality control to meet exacting specifications for wall chambers and variation.
Certifications: Conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Ordering information: Tubes for 800 and 900MHz NMR are also available, contact your Avantor sales representative for more information.
Disposable and thrift-grade tubes do not have caps attached, high-quality tubes are supplied with caps attached.