KIMBLE® Short Path Distillation Head, Vigreux, DWK Life Sciences
Supplier: DWK Life Sciences
287120-0000
KT287120-0000EA 783.47 CAD
KT287120-0000
KIMBLE® Short Path Distillation Head, Vigreux, DWK Life Sciences
Distillation Components Distillation Heads
Design uses Vigreux indentations for increased surface area within the vapor path. This promotes better refluxing prior to condensation, yielding a better separation.
With a standard taper thermometer in place, the bulb and stem serve as column packing. Closely coupled condenser and vacuum connections provide a short condensate travel path.
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.