COMPASS™ CX Compact Scales, Ohaus
Supplier: Ohaus
30467761 30467762 30428203 30467747 30428199 30428201 30428200 30467746 30428202
76377-592EA 422.93 CAD
76377-592 76377-594 76377-596 76377-598 76377-600 76377-602 76377-604 76377-606 76377-608
COMPASS™ CX Compact Scales, Ohaus
Balances and Scales
Designed with energy efficient weighing in mind for workplace and in-the-field weighing.
  • Streamlined two-button operation means that the balance can be operated easily
  • Large backlit LCD display enables easy viewing of weighing results even in dim lighting conditions, and from a distance
The COMPASS™ CX series features alkaline batteries capable of up to 1000 hours of continuous use, providing peace of mind for daily weighing applications.
The COMPASS™ CX comes in an ingenious space-saving design, which allows you to stack up to 4 scales to save storage space. Additionally, the CX features a standard stainless steel pan for durable use.
Applications include weighing, display hold.
Weighing units supported include Gram, Pound, Ounce, Newton. For CX221 and CX221P Pound's are not used.
Ordering information: A range of optional accessories are available.