KIMBLE® HI-VAC® Straight Valves, with Glass Plug and Tip O-rings, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
826450-0004 826450-0008 826450-0012
KT826450-0004CS 283.05 CAD
KT826450-0004 KT826450-0008 KT826450-0012
KIMBLE® HI-VAC® Straight Valves, with Glass Plug and Tip O-rings, DWK Life Sciences
Valves Vacuum Valves
This valve uses a glass plug (as an alternative to PTFE or CTFE) for applications where the 'out-gassing' plug material is undesirable.
  • Designed with a precision external thread for fine control
  • This valve can be used as a bleed up to ¹/₂ turn
  • Fully open position is at 1-¹/₂ turns
  • Greater solvent resistance
All plugs are totally interchangeable with other HI-VAC® counterparts.
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Delivery information: Supplied with FKM O-rings.