Analytical Balances, Entris® II Essential, Sartorius
Supplier: Sartorius
BCE124-1S BCE124I-1S BCE224-1S BCE224I-1S BCE64-1S BCE64I-1S
CA76333-832EA 3463.3 CAD
CA76333-832 CA76333-834 CA76333-848 CA76334-160 CA76334-198 CA76334-200
Analytical Balances, Entris® II Essential, Sartorius
Balances and Scales
These Entris® II laboratory balances are ideal for basic weighing tasks.
  • Highly accurate results guaranteed via Monolithic weigh cell technology
  • Fastest stabilization time in its class using state-of-the-art weighing sensors
  • Future-proof USB Type C interface and industry-proof RS232 9-pin interface
  • Prevents cross contamination with wipe clean design and easy-to-remove parts
  • isoCAL (internal calibration and adjustment) for total assurance of accurate weighing results
  • Best repeatability with rectangular weighing pan
  • Overload protection with rugged design
More Product Information