Polytron® Generator, PT-DA/BEC (BIOTRONA), Kinematica
Supplier: Kinematica
11030034 11030286 11030288 11030291
76360-428EA 4580.2 CAD
76360-428 76360-430 76360-432 76360-434
Polytron® Generator, PT-DA/BEC (BIOTRONA), Kinematica
Homogenizers Dispersing Tools
BIOTRONA® design high turbulence mixing. High-turbulence mixing head with low shear forces and energy inputs. Time saving with improved results compared to stirrers and dissolvers of up to 90%. Dispersing generators are made from high-alloy stainless steel 316L, electropolished, Ra≤1.6 μm as standard.
  • Fast dissolving and suspending of solid particles into liquids
  • Ideal for higher viscosities
  • Lower shear forces