Clarity FORENSIC Drugs of Abuse Urine Single Dip Test, Clarity Diagnostics
Supplier: Clarity Diagnostics
DSM-114 DET-114 FTN-114
76313-706BX 169.72 CAD
76313-706 76313-708 76313-710
Clarity FORENSIC Drugs of Abuse Urine Single Dip Test, Clarity Diagnostics
Drug Tests
The Clarity FORENSIC single dip test is a rapid test to detect Drugs of Abuse in urine sample in less than 5 minutes.
  • Dip and read test
  • For Forensic IVD use only
  • Detects up to 80 hours of alcohol abuse
  • 25 tests/box
  • Results can be photocopied
These tests are widely used for clinical trials, pre-employment and employee screening. They are currently classified as an FDA Exempt test.