Soxhlet Extraction Apparatus, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
416000 416005 415005 415010 334003 334014 334025 415000
80093-780CS 1238.34 CAD
80093-780 80093-782 80093-786 80093-788 80093-790 80093-792 80093-794 80093-784
Soxhlet Extraction Apparatus, DWK Life Sciences
Extractors Soxhlet and Soxhlet-Type Extractors
Soxhlet extraction apparatus with standard taper ground joints is used for continuous extraction of analytes from a solid into an organic solvent.
  • Manufactured from low extractable borosilicate glass
As the flask containing the solvent is heated, vapors rise in the larger outside tube, enter the water-cooled condenser and liquate and when the liquid level in the extractor reaches the top of the bent tube, siphoning action returns the extract-enriched solvent to the flask.
Certifications: Conforms to USP Type I and ASTM E 438 Type I, Class A requirements