WHEATON® Safety Coated Glass Bottles, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
W217850 W217951 W217952 W217953 W217954 W217955 W217851 W217852 W217853 W217854 W217855 W217856
10847-496CS 646.49 CAD
10847-496 10847-554 10847-556 10847-558 10847-560 10847-562 10847-564 10847-570 10847-572 10847-574 10847-576 10847-578 10847-580
WHEATON® Safety Coated Glass Bottles, DWK Life Sciences
Bottles Glass Bottles
Plastisol coats the exterior of the bottle to retain contents if a bottle is accidentally broken.
  • Made from clear or amber soda-lime glass that conforms to USP Type III requirements
  • Available with or without caps
  • 4, 8, 16, and 32 oz. Boston round style formats
These bottles are packaged in convenience packs with caps attached to maintain cleanliness.