VAI® SMS Garments, Veltek
Supplier: Veltek Associates
1600-LC-S-1648 1600-LC-S-1647 1600-LC-S-1645 1800P-E-S-1800 1800P-E-S-1801 1800P-E-S-1802 1800P-E-S-1803 1800P-E-S-1804 1800P-E-S-1805 1800P-E-S-1806 1800P-E-S-1807 1800P-EHB-S-1808 1800P-EHB-S-1809 1800P-EHB-S-1810 1800P-EHB-S-1811 1800P-EHB-S-1812 1800P-EHB-S-1813 1800P-EHB-S-1814 1800P-EB-S-1815 1800P-EB-S-1816 1800P-EB-S-1817 1800P-EB-S-1818 1800P-EB-S-1819 1800P-EB-S-1820 1800P-EB-S-1821 1800P-H-UC-S-1822 1800P-H-UC-S-1823 1800P-HM-S-1824 1800P-HM-S-1825
75795-424CS 494.54 CAD
75795-424 75795-534 75795-594 75795-596 75795-598 75795-650 76308-366 76308-368 76308-370 76308-372 76308-374 76308-376 76308-378 76308-380 76308-382 76308-384 76308-386 76308-388 76308-390 76308-392 76308-394 76308-396 76308-398 76308-400 76308-402 76308-404 76308-406 76308-408 76308-410 76308-412 76308-414 76308-416
VAI® SMS Garments, Veltek
Disposable Garments
The SMS garment line is constructed with a high quality spunbond-meltblown-spunbond (SMS) non-woven polypropylene fabric that provides excellent breathability while maintaining protection.
  • Highly breathable fabric releases heat while maintaining protection
  • Garments feature elastic in high movement areas for ease of movement and appropriate fit
  • 100% bound seam construction for increased durability and reduced particle permeability
  • Individually packaged in easy to tear bags with two liner bags per case
  • All packaging is labled with lot number and expiration
More Product Information