HyFlex® 74-050 Cut-Resistant Gloves, Ansell
Supplier: Ansell
HyFlex®
245326
76377-478CS 732.2 CAD
76377-478
HyFlex® 74-050 Cut-Resistant Gloves, Ansell
Gloves
A close, comfortable fit with the protection of a heavyweight glove.
  • Excellent levels of cut protection: ANSI Level A6
  • Incorporates Tuff-Cuff™ anti-shrink, extended cuff construction
  • Excellent protection in food processing applications
  • Good for most knife-hand and some off-hand applications
  • Available in white and gray
Offering a high level of cut protection and lightweight comfort, HyFlex® 74-050 series gloves provide superior dexterity and can be used as a liner under another glove.
Certifications: ANSI 3 Abrasion; ANSI A6 Cut; EN 388:2016 4X2XF; EN 388:2003 454X; EN 420:2003 + A1:2009.
Packaging: Gloves are packaged 12 pieces per case.