Autoclave-Resistant Labels for Laser Printers
Supplier: GA INTERNATIONAL INC
807656009476 807656009483 807656009490 807656009506 807656009513
76271-994PK 165.38 CAD
76271-994 76271-996 76271-998 76272-000 76272-002
Autoclave-Resistant Labels for Laser Printers
Labels
Laser labels feature a permanent adhesive, perfect for identifying disposable sterilization pouches and bags. Printable using most common desktop laser printers, they can be printed with human-readable text, serialized numbering, as well as 1D/2D barcodes.
  • Temperature range: −40 to +150 °C (−40 to +302 °F)
  • Strong permanent adhesive ensures labels adhere to glass, metal, and plastic surfaces
  • Designed for use in steam autoclaves, dry heat ovens and with gamma irradiators (up to 50 KGy), and ethylene oxide sterilizers
  • Compatible with all major brands of desktop laser printers
Permanent laser printable labels, ideal for glassware, surgical instruments, metal and plastic surfaces, trays, blister packs, and other items intended for use in steam autoclaves, ovens, and gamma irradiators. These autoclave laser labels are durable, inert, weather- and water-proof, alcohol-resistant, and have been tested to endure up to 20 autoclave cycles (at 121 °C/250 °F, 16.5 psi for 20 minutes). Compatible with all major brands of desktop laser printers.