KIMBLE® ACCUFORM® Graduated High Recovery Vials, Borosilicate, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
60700-1 60700-2 60700-5 60700-10 60700-3 60705-1 60705-5
66020-800PK 408.51 CAD
66020-800 66020-801 66020-804 66020-806 66020-904 66020-802 66020-902 66020-903 66020-905
KIMBLE® ACCUFORM® Graduated High Recovery Vials, Borosilicate, DWK Life Sciences
Vials
These ACCUFORM® vials are internally contoured for optimal performance with limited samples.
  • Graduated to ensure more accurate measurements and dispensing
  • Tapered bases allow for maximum sample recovery
  • Open-top closure and PTFE-faced silicone septa, assembled and attached
Clear glass is manufactured from 33 expansion, low extractable borosilicate glass. Amber glass is manufactured from 51 expansion borosilicate amber glass.
Certifications: Clear glass conforming to USP Type I and ASTM E438, Type I, Class A requirements. Amber glass conforming to USP, Type I requirements.