KIMBLE® Vacuum Hydrolysis Tubes, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
896860-2860 896860-8910
KT896860-4010EA 509.47 CAD
KT896860-4010 KT896860-2860 KT896860-4015 KT896860-8910
KIMBLE® Vacuum Hydrolysis Tubes, DWK Life Sciences
Tubes Test Tubes
Versatile tube for use where a vacuum environment is desired.
  • Useful in protein hydrolysis and amino acid separations
  • Other applications include sealed tube reactions, freeze drying and concentration
Manufactured from 33 expansion, low extractable borosilicate glass.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, class A requirements.
Caution: Not recommended for use over 150 °C.