KIMAX® KimCote® Round-Bottom Boiling Flasks, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
KC25330-100 KC25330-250 KC25330-1000 KC25330-2000
89097-400CS 175.56 CAD
89097-400 89097-402 89097-404 89097-406 89097-408
KIMAX® KimCote® Round-Bottom Boiling Flasks, DWK Life Sciences
Flasks Round Bottom Flasks
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
  • Spherical socket joint
  • Autoclavable
  • KimCote® safety coating reduces the hazards of shattered glass and leakage of toxic or corrosive chemicals
KimCote® boiling flasks with a round bottom, plastic-coated single neck flask useful as a rotary evaporator condensate collection flask or as a replacement for European designation KS 20/35.