Round Bottom Flasks, Single Standard Taper Neck, Heavy Wall, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
294010-0005 294010-0050 294010-0100 294010-0250 294010-0010 294010-0015 294010-0025 294010-0500
KT294010-0005EA 86.64 CAD
KT294010-0005 KT294010-0050 KT294010-0100 KT294010-0250 KT294010-0010 KT294010-0015 KT294010-0025 KT294010-0500
Round Bottom Flasks, Single Standard Taper Neck, Heavy Wall, DWK Life Sciences
Flasks Round Bottom Flasks
These are single neck flask with a standard taper outer joint.
  • Heavy wall
  • ASTM Method D95
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
SPECIFICATIONS
Environmentally Preferable