Vacuum Filtration Luer, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
205100-1420 205100-2440
93001-856EA 244.38 CAD
93001-856 93001-860
Vacuum Filtration Luer, DWK Life Sciences
Adapters Adapters, Joints with Tube Connectors
Use this adapter with disposable syringe cartridges for easy sample concentration and to eliminate the cleaning of glass frits and funnels. Designed to reduce cross-contamination.
  • Features top luer fitting and integral vacuum connection
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Ordering information: Includes drip tip. Plastic valves with luer fittings are available separately.