Wizard Series PVC/Nylon/PVC 60" Coats with Detachable Hood, MCR Safety
Supplier: MCR Safety
360CX2 360CM 360CL 360CXL 360CX3 360CX4 360CX6 360CX5 360CX7
76341-308EA 131.38 CAD
76341-308 76341-430 76341-520 76341-620 76341-696 76341-782 76341-818 76341-844 76341-862
Wizard Series PVC/Nylon/PVC 60" Coats with Detachable Hood, MCR Safety
Workwear
This wizard series coat features a 0.28 mm double coating of PVC, with tough nylon scrim material layered in between.
  • 60" ankle length design
  • Detachable drawstring hood
  • Snaps on front and back can fit together to form chaps
  • Rip stop scrim material provides excellent resistance to snags and tears
Nylon scrim has excellent rip and puncture resistance. This yellow two-piece coat is a 60" (ankle length) option that features a detachable drawstring hood and storm flap under a snap up front closure. Snaps on the lower front and back can fit together to form 'chaps' around each leg. Limited flammability, this garment is not intended for use as protection against ARC flash or flash fire.