Wizard Series 3-Piece Suit with Hood, Rain Jacket, and Bib Pants, MCR Safety
Supplier: MCR Safety
3003S 3003M 3003X3 3003L 3003XL 3003X2 3003X4 3003X5
76341-406EA 95.4 CAD
76341-406 76341-542 76341-648 76341-664 76341-744 76341-796 76341-966 76341-980
Wizard Series 3-Piece Suit with Hood, Rain Jacket, and Bib Pants, MCR Safety
Workwear
The wizard series material provides a 0.28 mm double coating of PVC with tough nylon scrim material layered in between.
  • Three-piece suit includes jacket with detachable drawstring hood and bib pants
  • Take up snaps on the wrists and ankles
  • Snap front jacket, bib-style pants
  • PVC/nylon/PVC construction
The nylon scrim has excellent rip and puncture resistance. The jacket features a storm fly front with snaps, take up snaps at the wrists, and a detachable hood with drawstring. Bib pants feature a fly front with snap and take up snaps at the waist and ankles. Limited flammability not intended for use as protection against ARC flash or flash fire.