DURAN® SUPER DUTY Erlenmeyer Flask, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
212173606 212273604
76186-136CS 535.48 CAD
76186-136 76193-154
DURAN® SUPER DUTY Erlenmeyer Flask, DWK Life Sciences
Flasks Erlenmeyer Flasks
The DURAN® SUPER DUTY products are characterized by a higher mechanical strength achieved by reinforcing the rim.
  • Autoclavable
  • Retrace code
Application note: To avoid breakages due to thermal stress, uniform and slow heating of SUPER DUTY products is recommended.
Order Now
SPECIFICATIONS
Environmentally Preferable