Chromatography Paper
Supplier: United Scientific Supplies
CHRP050
470320-990PK 6.6 CAD
470320-990
Chromatography Paper
Filters Filter Papers Chromatography Paper
Separate the different colors of a solution.
  • Dimension: 12.5×1.75 cm
Chromatography paper is used to separate colored chemicals. This is done by taking advantage of the different rates of migration across paper.
Packaging: Sold as a pack of 50 strips.