Black Phenolic Screw Caps, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
240683 239253
53499-472CS 4412.92 CAD
53499-472 10796-536
Black Phenolic Screw Caps, DWK Life Sciences
Closures Bottle Closures
The unique cone liner design offers superior torque retention and excellent sealing characteristics.
  • 14B Rubber lined caps feature a nontoxic white styrene-butadiene rubber liner ideal for cell culture work
  • Aluminum foil faced pulp board liners can be used with organic solvents
  • Use low density polyethylene (LDPE) lined caps with distilled water, analytical standards, and reagents
  • Open top caps with gray chlorobutyl/50 septa are ideal for use with WHEATON® media bottles
  • Open top caps provide access with a syringe
Manufactured from LDPE. The unique cone design provides a wedge-type seal that not only seals across the top but also across the inside diameter.