Chromolith® WidePore (WP) 300, HPLC Columns for Bioapplications, MilliporeSigma
Supplier: MilliporeSigma
1.52265.0001 1.52266.0001 1.52267.0001 1.52268.0001 1.52270.0001 1.52271.0001 1.52251.0001 1.52252.0001 1.52254.0001 1.52272.0001 1.52273.0001 1.52250.0001
CA76053-600EA 1673 CAD
CA76053-600 CA76053-602 CA76053-604 CA76053-606 CA76053-608 CA76053-610 CA76053-612 CA76053-614 CA76053-616 CA76053-620 CA76053-622 CA76053-624
Chromolith® WidePore (WP) 300, HPLC Columns for Bioapplications, MilliporeSigma
Chromatography Columns
These columns have been designed following the high demand for suitable analytical methods for process monitoring and quality control of wide pore biomolecules. Accurate analysis of proteins, antibodies and large peptides requires columns with good permeability, along with better mass transfer and selectivity. In order for size exclusion not to influence the separation, the pore size should be approximately ten times larger than the molecule being analyzed.
  • Completely bioinert column hardware
  • High bio-recovery
  • Selectivity for a range of biomolecules
  • Very low column backpressure
  • High resistance to column blockage
  • Cost savings from higher sample throughput and column durability
  • Possibility to use flow gradients
Wide pore (300 Å) monolithic silica columns are made of a single continuous bed rod of high purity porous silica that is then bonded with C18, C8, C4, epoxy and Protein A depending on the use of the column.