DURAN® Laboratory Bottles, Pressure Plus+, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
218152402 1175925 1175926 218155408
75995-264CS 1997.46 CAD
75995-264 75995-272 76204-552 76204-554
DURAN® Laboratory Bottles, Pressure Plus+, DWK Life Sciences
Bottles Glass Bottles
With easy-to-read scale and large labelling field for easy marking, in fired-on, highly durable ceramic.
  • Plastic coated
  • GL 45 screw thread
  • Autoclavable
  • Retrace Code
Pressure resistance conforms to DIN EN 1595, confirmed by GS marking (TÜV ID: 0000020716). Vacuum and/or pressure resistant from – 1 to 1.5 bar due to modified geometry (based on ISO 4796-1). When pressure loaded the following apply: thermal shock resistance 30 K and maximum usage temperature 140 °C.
Blue scale for visual differentiation from the standard laboratory bottle. The coating provides scratch, leak and splinter protection and is ideally suited to both the transport and storage of hazardous media or valuable samples. Typical applications: safe working under pressure or vacuum, sampling under pressure, storage of gas generating media.