Self-Laminating Calibration Labels
Supplier: GA INTERNATIONAL INC
807656032955 807656040387 807656110936 807656040479 807656118734
76210-160RL 145.36 CAD
76210-160 76275-528 76275-530 76275-534 76275-546 76275-548 76275-550 76275-552
Self-Laminating Calibration Labels
Labels
Self-laminating calibration labels for secure and safe identification of regularly inspected equipment and instruments. These labels can be inscribed with all relevant information and then laminated with the supplied clear laminate to provide physical protection against various abuses.
  • Temperature range: −80 to +93 °C (−112 to +200 °F)
  • Can withstand extreme temperatures, including cryogenic and high heat conditions
  • Resists abrasion and exposure to harsh chemicals and alcohols
  • Ideal for use with equipment that will undergo preventative maintenance,
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable