Single Channel Pipettes, Variable Volume, Labsciences
Supplier: LAB SCIENCE CA
5000-VV 10000-VV
CA11020-004EA 385.55 CAD
CA11020-004 CA11020-006
Single Channel Pipettes, Variable Volume, Labsciences
Pipettes Micropipettes
These pipettes have an ergonomic, lightweight design with a soft plunger movement to minimize hand fatigue and RSI.
  • Simply turn the plunger for quick and easy volume setting
  • Click stop digital adjustable volume mechanism
  • Semi-autoclavable
  • Uses Industry Standard pipette tips
  • Color coded by volumes for easy identification
  • Two stage plunger operation for 'blow-out' step during pipeting
The streamlined tip ejector allows easy access to narrow containers/tubes.
Certifications: Individually calibrated in accordance with DIN 12650 Standards and suppliedMore Product Information