Single Channel Pipettes, Fixed Volume, Labsciences
Supplier: LAB SCIENCE CA
1000-FV 10000-FV 2000-FV
CA11019-998EA 342.47 CAD
CA11019-998 CA11020-002 CA11027-118
Single Channel Pipettes, Fixed Volume, Labsciences
Pipettes Micropipettes
These pipettes have an ergonomic, lightweight design with a soft plunger movement to minimize hand fatigue and RSI.
  • Semi-autoclavable
  • Uses Industry Standard pipette tips
  • Color coded by volumes for easy identification
  • Two stage plunger operation for 'blow-out' step during pipetting
The streamlined tip ejector allows easy access to narrow containers/tubes.
Certifications: Individually calibrated in accordance with DIN 12650 Standards and supplied with a computer generated calibration report.
Delivery information: Includes a mountable pipette holder for easy storage and a calibration tool for easy calibration and service.