Cryogenic and Autoclave-Resistant Self-Laminating Laser Labels
Supplier: GA INTERNATIONAL INC
807656009438 807656009445 807656009452 807656028767 807656028774 807656028750 807656009469
76271-702PK 117.58 CAD
76271-702 76271-704 76271-708 76271-982 76271-984 76271-986 76271-988 76271-990 76271-992
Cryogenic and Autoclave-Resistant Self-Laminating Laser Labels
Labels
These wrap-around labels for laser printers withstand long-term cryo storage in liquid nitrogen and deep-freeze conditions, as well as sterilization in steam autoclaves and boiling water.
  • "Temperature range: −196 to +150 °C (−321 to +302 °F)
  • Transparent laminating tail provides extra protection against scratching and spraying with chemicals and detergents
  • Printable with all major desktop laser printers
  • Withstands extreme elevated temperatures (150 °C), able to resist steam autoclaves and boiling water
  • Resistant to gamma irradiation (tested up to 50 KGy)
Cryogenic wrap-around laser labels for long-term cryo storage in liquid nitrogen and deep freeze harsh environments. Resistant to autoclave, boiling water, as well as spraying with ethanol, IPA, acetone, and other chemicals and detergents. Cryo labels accept desktop laser printing and permanent marker. Water-resistant, wear- and tear-proof, labels will not jam printers and are resistant to UV and gamma irradiation.