Tamper-Evident Cryogenic Labels
Supplier: GA INTERNATIONAL INC
807656082714 807656082721 807656082738 807656082745 807656082752 807656082769
76273-942RL 106.22 CAD
76273-942 76273-944 76273-946 76273-948 76273-950 76273-952 76273-954 76273-956 76273-962 76273-964 76273-966 76273-968 76273-970 76273-972
Tamper-Evident Cryogenic Labels
Labels
Tamper-evident cryogenic labels will tear upon any attempt to remove them. Perfect for the secure storage and transportation of valuable or sensitive samples.
  • Temperature range: −196 to +110 °C (−321 to +230 °F)
  • Tamper-evident seal ensures any attempt to open the container will be noticed
  • Able to withstand immersion in liquid nitrogen and deep-freeze storage conditions
  • Printable with all major thermal transfer printers
Thermoplastic, waterproof cryogenic labels will tear upon any attempt to remove them. Ideal for cryogenic vials, cryo boxes, and other containers that require tamper-evidence when stored in liquid nitrogen, dry ice, and freezers. Advance and practical design adds rigidity and strength to reduce accidental seal ruptures, yet destructs easily upon removal. Printable with thermal transfer barcode printers.