High Vacuum Manifolds, Solid Glass Stopcocks, ChemScience, SATI International Scientific
Supplier: CHEM SCIENCE INC CA
CS-M0230005 CS-M0240003 CS-M0240004 CS-M0240005 CS-M0260003 CS-M0260004 CS-M0260005 CS-M0280003 CS-M0280004 CS-M0280005 CS-M0290003 CS-M0290004 CS-M0290005 CS-M0300003 CS-M0300004 CS-M0300005 CS-M0250003 CS-M0250004 CS-M0250005 CS-M0230003 CS-M0230004
CA10202-626CS 1855.67 CAD
CA10202-626 CA10202-628 CA10202-630 CA10202-632 CA10202-634 CA10202-636 CA10202-638 CA10202-640 CA10202-642 CA10202-644 CA10202-646 CA10202-648 CA10202-650 CA10202-652 CA10202-654 CA10202-656 CA10204-478 CA10204-480 CA10204-482 CA10204-484 CA10204-486
High Vacuum Manifolds, Solid Glass Stopcocks, ChemScience, SATI International Scientific
Manifolds
These manifolds are delivered with 4 mm double oblique, high vacuum solid glass stopcocks precision ground with corresponding numbers on barrel and plug.
  • Beams have 22 mm O.D. with 10 mm hose connections
  • Distance between stopcocks is 100 mm from center to center
Solid glass stopcocks have constant pressure retention devices.