Vacuum Manifolds, Inert Gas, Double, All Glass, Chemscience, SATI International Scientific
Supplier: CHEM SCIENCE INC CA
CS-M0411003 CS-M0411004 CS-M0411005 CS-M0181003 CS-M0181004 CS-M0181005 CS-M0401003 CS-M0401004 CS-M0401005 CS-M0421003 CS-M0421004 CS-M0421005 CS-M0431003 CS-M0431004 CS-M0431005 CS-M0441003 CS-M0441004 CS-M0441005
CA10202-590CS 1178.66 CAD
CA10202-590 CA10202-592 CA10202-594 CA10202-596 CA10202-598 CA10202-600 CA10202-602 CA10202-604 CA10202-606 CA10202-608 CA10202-610 CA10202-612 CA10202-614 CA10202-616 CA10202-618 CA10202-620 CA10202-622 CA10202-624
Vacuum Manifolds, Inert Gas, Double, All Glass, Chemscience, SATI International Scientific
Manifolds
These manifolds are delivered with 4 mm double oblique, high vacuum hollow glass stopcocks precision ground with corresponding numbers on barrel and plug.
  • Beams have 22 mm O.D. with 10 mm hose connections
  • Distance between stopcocks is 100 mm from center to center
Lower vacuum chambers on stopcock are evacuated with a separate hole in the plug in order to hold the plug in the stopcock barrels.