Vacuum Manifolds, Inert Gas, Double, ChemScience, SATI International Scientific
Supplier: CHEM SCIENCE INC CA
CS-M0111003 CS-M0111004 CS-M0111005 CS-M0171003 CS-M0171004 CS-M0171005 CS-M0141003 CS-M0141004 CS-M0141005 CS-M0101003 CS-M0101004 CS-M0101005 CS-M0471003 CS-M0471004 CS-M0471005
CA10202-560CS 1500.4 CAD
CA10202-560 CA10202-562 CA10202-564 CA10202-566 CA10202-568 CA10202-570 CA10202-572 CA10202-574 CA10202-576 CA10202-578 CA10202-580 CA10202-582 CA10202-584 CA10202-586 CA10202-588
Vacuum Manifolds, Inert Gas, Double, ChemScience, SATI International Scientific
Manifolds
General purpose double bank manifold for use with vacuum and inert gas.
  • Both banks are made from 22 mm medium wall tubing and have all banks open
  • Ports are spaced 100 mm center to center and have an O.D. of 10 mm
Delivery information: Supplied with 0 to 4 mm high vacuum valves for grease-free operation.