Valves, High Vacuum, Protected, Low Hold Up, Metering, PTFE , Chemscience
Supplier: CHEM SCIENCE INC CA
CS-S0680004 CS-S0680008 CS-S0680012
CA10203-134CS 138.04 CAD
CA10203-134 CA10203-136 CA10203-138
Valves, High Vacuum, Protected, Low Hold Up, Metering, PTFE , Chemscience
Valves Vacuum Valves
Viton® O-rings on PTFE shaft are protected with precision PTFE wipers.
  • With two 9 mm O.D. side arms bent to minimize hold up of liquids
This design provides for an excellent vacuum tight seal without exposing any material other than PTFE to chemicals.