Valves, High Vacuum, Protected, Three Way, PTFE , Chemscience
Supplier: CHEM SCIENCE INC CA
CS-S0640004 CS-S0640008 CS-S0640012
CA10203-122CS 343.94 CAD
CA10203-122 CA10203-124 CA10203-126
Valves, High Vacuum, Protected, Three Way, PTFE , Chemscience
Valves Vacuum Valves
Viton® O-rings on PTFE shaft are protected with precision PTFE wipers.
  • This design provides an excellent vacuum tight seal
  • Does not expose any material other than PTFE to chemicals
The plug has a machined-in integral PTFE tip O-ring to make the primary seal.