Valves, High Vacuum, Protected, Low Hold Up, PTFE, SATI International Scientific
Supplier: CHEM SCIENCE INC CA
CS-S0630004 CS-S0630008 CS-S0630012
CA10203-116CS 187.52 CAD
CA10203-116 CA10203-118 CA10203-120
Valves, High Vacuum, Protected, Low Hold Up, PTFE, SATI International Scientific
Valves Vacuum Valves
With two 9 mm O.D. sidearms bent to minimize hold up of liquids.
  • Viton® O-rings on PTFE shaft are protected with precision PTFE wipers
  • The plug has a machined-in integral PTFE tip O-ring to make the primary seal
This design provides for an excellent vacuum tight seal without exposing any material other than PTFE to chemicals.