Valves, High Vacuum, Protected, 180°, PTFE , Chemscience
Supplier: CHEM SCIENCE INC CA
CS-S0620012 CS-S0620004 CS-S0620008
CA10203-114CS 206.49 CAD
CA10203-114 CA10204-524 CA10204-526
Valves, High Vacuum, Protected, 180°, PTFE , Chemscience
Valves Vacuum Valves
The PTFE plug has a machined-in integral PTFE tip O-ring to make the primary seal.
  • With two 9 mm O.D. side arms sealed to the barrel at a 180° angle to one another
  • Viton® O-rings on PTFE shaft are protected with precision PTFE wipers
This design provides for an excellent vacuum tight seal without exposing any material other than PTFE to chemicals.