Valves, High Vacuum, 180°, O-Ring, PTFE, Chemscience
Supplier: CHEM SCIENCE INC CA
CS-S0560004 CS-S0560008 CS-S0560012
CA10203-106CS 158.19 CAD
CA10203-106 CA10203-108 CA10203-110
Valves, High Vacuum, 180°, O-Ring, PTFE, Chemscience
Valves Vacuum Valves
With two 9 mm O.D. side arms sealed to the barrel at a 90° angle to one another.
  • The non-rotating PTFE plug has three O-rings
  • The tip O-ring making the primary seal on the precision formed tapered seat
For replacement Viton® O-ring kit, see CS-R7010 or CS-R108110 or CS-R110112.