Plugs, High Vacuum Valve, PTFE Stem, Viton O-Ring, Chemscience
Supplier: CHEM SCIENCE INC CA
CS-S0540004 CS-S0540008 CS-S0540012
CA10203-094CS 76.65 CAD
CA10203-094 CA10203-096 CA10203-098
Plugs, High Vacuum Valve, PTFE Stem, Viton O-Ring, Chemscience
Valves Vacuum Valves
The plug is complete with control knob and PTFE stem having three O-rings.