Vacuum Traps, Serrated Hose Connections , Chemscience
Supplier: CHEM SCIENCE INC CA
CS-V00163229 CS-V00164050 CS-V00165050
CA10203-054CS 216.38 CAD
CA10203-054 CA10203-058 CA10203-062
Vacuum Traps, Serrated Hose Connections , Chemscience
Vacuum Traps
With joints for easy removal of condensate.
  • Serrated hose connections for side tube
  • O.D. of 13 mm at the smallest serration
  • O.D. of 16 mm at the largest serration
Center tube hose connection is always 10 mm for easy connection to manifold bank tubes which are also with 10 mm hose connection.